Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112176 Supplier Invitrogen™ Supplier No. PA5112176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KBP Polyclonal Antibody for Western Blot, IHC (P)

Required for neuronal development and differentiation. Required for organization of axonal microtubules, and axonal outgrowth and maintenance during peripheral and central nervous system development. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KBP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene KIFBP
Gene Accession No. Q96EK5
Gene Alias 0710007C18Rik; 2510003E04Rik; hypothetical protein LOC527433; hypothetical protein LOC606294; Kbp; Kiaa1279; KIF1 binding protein; KIF1-binding protein; KIF1-binding protein-like protein; Kif1bp; KIF-binding protein; KIFBP; kinesin binding protein; kinesin family binding protein; LOC606294; mKIAA1279; TTC20
Gene Symbols KIFBP
Host Species Rabbit
Immunogen E. coli-derived Human KIAA1279/KIF1BP fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26128
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.