Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KBP Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501989286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9286 Supplier Bioss Supplier No. BS9716R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a kinesin family member 1 binding protein that is characterized by two tetratrico peptide repeats. The encoded protein localizes to the mitochondria and may be involved in regulating transport of the mitochondria. Mutations in this gene are associated with Goldberg-Shprintzen megacolon syndrome.

Specifications

Antigen KBP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KIF1BP
Gene Accession No. Q6ZPU9
Gene Alias 0710007C18Rik; 2510003E04Rik; BOS_24207; KBP; Kiaa1279; KIAA1279 ortholog; KIF1 binding protein; KIF1-binding protein; Kif1bp; KIF-binding protein; Kifbp; LOC100346136; mKIAA1279
Gene Symbols KIF1BP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KBP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72320
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.