Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KBTBD10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | KBTBD10 |
---|---|
Concentration | 0.35 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4966
![]() ![]() |
Proteintech Group Inc
255771AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
KLHL41 is involved in skeletal muscle development and differentiation. It regulates proliferation and differentiation of myoblasts and plays a role in myofibril assembly by promoting lateral fusion of adjacent thin fibrils into mature, wide myofibrils. It is required for pseudopod elongation in transformed cells.Specifications
KBTBD10 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
A2AUC9, O60662 | |
10324, 228003 | |
KBTBD10 Fusion Protein Ag22311 | |
Primary | |
-20°C | |
KLHL41 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KBTBD10, Kel like protein 23, Kelch related protein 1, KRP1, SARCOSIN | |
KLHL41 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title