Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KBTBD11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546490 Supplier Invitrogen™ Supplier No. PA546490
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GGPTGLQPFR CAALDGAIYC VSRAGTWRFQ PAREGEAGGD AGQGGGFEAL Sequence homology: Human: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KBTBD11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KBTBD11
Gene Accession No. O94819
Gene Alias 2900016B01Rik; 4930465M17Rik; Chronic myelogenous leukemia-associated protein; CMLAP; Kbtbd11; Kelch domain-containing protein 7B; kelch repeat and BTB (POZ) domain containing 11; kelch repeat and BTB domain containing 11; kelch repeat and BTB domain-containing protein 11; KIAA0711; KLHDC7C; mKIAA0711
Gene Symbols KBTBD11
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human KBTBD11 (aa 545-594).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9920
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.