Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KBTBD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598599
Description
KBTBD2 Polyclonal Antibody for Western Blot, ELISA
The KBTBD2 gene encodes the Kelch repeat and BTB (POZ) domain-containing protein 2, which is involved in various cellular processes, including protein ubiquitination and metabolic regulation. The KBTBD2 protein functions as a substrate adaptor for the Cullin-RING E3 ubiquitin ligase complex, facilitating the ubiquitination and subsequent proteasomal degradation of specific substrate proteins. This activity is crucial for maintaining cellular protein homeostasis and regulating metabolic pathways. KBTBD2 has been implicated in glucose metabolism and insulin signaling, where it plays a role in modulating the degradation of key regulatory proteins. Alterations in KBTBD2 function or expression are associated with metabolic disorders, such as obesity and type 2 diabetes. Additionally, KBTBD2's involvement in ubiquitin-mediated proteolysis underscores its significance in controlling various cellular responses and adaptations, making it a potential target for therapeutic interventions in metabolic diseases.
Specifications
KBTBD2 | |
Polyclonal | |
Unconjugated | |
KBTBD2 | |
BC022962; BKLHD1; BTB and kelch domain containing 1; BTB and kelch domain-containing protein 1; CGI-73; im:7146937; KBTBD2; kelch repeat and BTB (POZ) domain containing 2; kelch repeat and BTB domain containing 2; kelch repeat and BTB domain-containing protein 2; Kelch repeat and BTB domain-containing protein 2-like protein; KIAA1489; mKIAA1489; zgc:152954 | |
Rabbit | |
Protein G | |
RUO | |
25948 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8IY47 | |
KBTBD2 | |
Recombinant Human Kelch repeat and BTB domain-containing protein 2 protein (182-406AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction