Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KBTBD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532116
Description
Recommended positive controls: PC-3. Predicted reactivity: Mouse (89%), Rat (88%), Rhesus Monkey (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The BTB (Broad-Complex, Tram track and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. KBTBD3 (kelch repeat and BTB domain-containing protein 3), also known as BKLHD3, is a 608 amino acid protein that contains one BACK (BTB/Kelch associated) domain, one BTB (POZ) domain and five kelch repeats. The gene encoding KBTBD3 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
Specifications
KBTBD3 | |
Polyclonal | |
Unconjugated | |
KBTBD3 | |
2200003A07Rik; Bklhd3; BTB and kelch domain containing 3; BTB and kelch domain-containing protein 3; Kbtbd3; kelch repeat and BTB (POZ) domain containing 3; kelch repeat and BTB domain containing 3; kelch repeat and BTB domain-containing protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
143879 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NAB2 | |
KBTBD3 | |
Recombinant protein fragment corresponding to a region within amino acids 11 and 268 of Human KBTBD3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction