Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KBTBD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561495
Description
Immunogen sequence: IMDAIKCVQG SGGLFPDARP STTEKYIFIH KTEENGENQY TFCYNIKSDS WKILPQSHLI DLPGSSLSSY GEKIFLTGGC KGKCCRTVRL HIAESYHDAT DQTWCY Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
The BTB (Broad-Complex, Tram track and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. KBTBD3 (kelch repeat and BTB domain-containing protein 3), also known as BKLHD3, is a 608 amino acid protein that contains one BACK (BTB/Kelch associated) domain, one BTB (POZ) domain and five kelch repeats. The gene encoding KBTBD3 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
Specifications
KBTBD3 | |
Polyclonal | |
Unconjugated | |
KBTBD3 | |
2200003A07Rik; Bklhd3; BTB and kelch domain containing 3; BTB and kelch domain-containing protein 3; Kbtbd3; kelch repeat and BTB (POZ) domain containing 3; kelch repeat and BTB domain containing 3; kelch repeat and BTB domain-containing protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
143879 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NAB2 | |
KBTBD3 | |
Recombinant protein corresponding to Human KBTBD3. Recombinant protein control fragment (Product #RP-97514). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction