Learn More
Invitrogen™ KBTBD3 Polyclonal Antibody

Description
Immunogen sequence: GSQDIKSLLD VESYNPLSRE WTSVSPLPRG IYYPEASACQ NIIYVLGSEV EIADAFNPSL DCFFKYNATT DQWSELVAEF GQFFHATLIK AVPVNCTLYI CDLSTYKVYS FCPDTCVWKG EGSFECAGFN AGAIGIEDKI YILGGDYAPD EITDEVQVYH SSRSEWEEVS PMPRALTEFY CQVIQFNKYR DPWYSNHF; Positive Samples: PC-3.
Specifications
Specifications
| Antigen | KBTBD3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.48 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | KBTBD3 |
| Gene Accession No. | Q8NAB2 |
| Gene Alias | 2200003A07Rik; Bklhd3; BTB and kelch domain containing 3; BTB and kelch domain-containing protein 3; Kbtbd3; kelch repeat and BTB (POZ) domain containing 3; kelch repeat and BTB domain containing 3; kelch repeat and BTB domain-containing protein 3 |
| Gene Symbols | KBTBD3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.