Learn More
Invitrogen™ KBTBD4 Polyclonal Antibody

Description
Recommended positive controls: U87-MG, SK-N-SH, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (100%), Chicken (92%), Rhesus Monkey (99%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | KBTBD4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.73 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | KBTBD4 |
| Gene Accession No. | Q8R179, Q9NVX7 |
| Gene Alias | 2510026C23Rik; BKLHD4; BTB and kelch domain containing 4; BTB and kelch domain-containing protein 4; HSPC252; KBTBD4; kelch repeat and BTB (POZ) domain containing 4; kelch repeat and BTB domain containing 4; kelch repeat and BTB domain-containing protein 4 |
| Gene Symbols | KBTBD4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.