Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KBTBD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555085
Description
Immunogen sequence: LESRVERHPL VRAQPELLRK VQMVKDAHEG RITTLRKKKK GKDGAGAKEA DKGTSKAKAE EDEEAERILP GILNDTLRFG MFL Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
The exact function of KBTBD5 remains unknown.
Specifications
KBTBD5 | |
Polyclonal | |
Unconjugated | |
Klhl40 | |
2310024D23Rik; KBTBD5; kelch like family member 40; kelch repeat and BTB (POZ) domain containing 5; kelch repeat and BTB domain-containing protein 5; kelch-like 40; kelch-like family member 40; kelch-like protein 40; kelch-like protein 40; LOW QUALITY PROTEIN: kelch-like protein 40; Klhl40; NEM8; nemaline myopathy type 8; sarcosynapsin; SRYP; SYRP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
131377 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2TBA0 | |
Klhl40 | |
Recombinant protein corresponding to Human KBTBD5. Recombinant protein control fragment (Product #RP-93588). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction