Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KBTBD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562568 Supplier Invitrogen™ Supplier No. PA562568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDLIFMISEE GAVAYDPAAN ECYCASLSSQ VPKNHVSLVT KENQVFVAGG LFYNEDNKED PMSAYFLQFD HLDSEWL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

The exact function of KBTBD5 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KBTBD5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Klhl40
Gene Accession No. Q2TBA0
Gene Alias 2310024D23Rik; KBTBD5; kelch like family member 40; kelch repeat and BTB (POZ) domain containing 5; kelch repeat and BTB domain-containing protein 5; kelch-like 40; kelch-like family member 40; kelch-like protein 40; kelch-like protein 40; LOW QUALITY PROTEIN: kelch-like protein 40; Klhl40; NEM8; nemaline myopathy type 8; sarcosynapsin; SRYP; SYRP
Gene Symbols Klhl40
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KBTBD5. Recombinant protein control fragment (Product #RP-103117).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131377
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.