Learn More
Invitrogen™ KBTBD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113673
Description
Immunogen sequence: ELVPTELNDI WRYNEEEKKW EGVLREIAYA AGATFLPVRL NVLCLTKM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 100%; Rat: 79%.
The exact function of KBTBD5 remains unknown.
Specifications
KBTBD5 | |
Polyclonal | |
Unconjugated | |
Klhl40 | |
2310024D23Rik; KBTBD5; kelch like family member 40; kelch repeat and BTB (POZ) domain containing 5; kelch repeat and BTB domain-containing protein 5; kelch-like 40; kelch-like family member 40; kelch-like protein 40; kelch-like protein 40; LOW QUALITY PROTEIN: kelch-like protein 40; Klhl40; NEM8; nemaline myopathy type 8; sarcosynapsin; SRYP; SYRP | |
Rabbit | |
Affinity chromatography | |
RUO | |
131377 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q2TBA0 | |
Klhl40 | |
Synthetic peptide directed towards the C terminal region of human KBTBD5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.