Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KBTBD7 Antibody (OTI1G3), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272268UV
Description
KBTBD7 Monoclonal specifically detects KBTBD7 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
KBTBD7 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KBTBD7 | |
Full length recombinant protein of human KBTBD7 (NP_115514) produced in HEK293T cells. | |
0.1 mL | |
84078.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1G3 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
DKFZP434E2318, kelch repeat and BTB (POZ) domain containing 7, kelch repeat and BTB domain-containing protein 7 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction