Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KBTBD7 Monoclonal Antibody (OTI1G3)
GREENER_CHOICE

Catalog No. PIMA525171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525171 Supplier Invitrogen™ Supplier No. MA525171
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KBTBD7 Monoclonal Antibody for Western Blot, ICC/IF

The protein encoded by this gene is a transcriptional activator, having been shown to increase the transcription of activator protein-1 and serum response element. The encoded protein can also form a complex with KBTBD6 and CUL3, which regulates the ubiquitylation and degradation of TIAM1, which is a regulator of RAC1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KBTBD7
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1G3
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KBTBD7
Gene Accession No. Q8WVZ9
Gene Alias KBTBD7; kelch repeat and BTB (POZ) domain containing 7; kelch repeat and BTB domain containing 7; kelch repeat and BTB domain-containing protein 7; kelch repeat and BTB domain-containing protein 7-like
Gene Symbols KBTBD7
Host Species Mouse
Immunogen Full length human recombinant protein of KBTBD7 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100909827, 485467, 84078
Target Species Human, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.