Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542444 Supplier Invitrogen™ Supplier No. PA542444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MPHFTVVPVD GPRRGDYDNL EGLSWVDYGE RAELDDSDGH GNHRESSPFL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%.

SLC12A4 mediates electroneutral potassium-chloride cotransport when activated by cell swelling. SLC12A4 may contribute to cell volume homeostasis in single cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC12A4
Gene Accession No. Q9UP95
Gene Alias AW546649; CTC-479C5.17; Electroneutral potassium-chloride cotransporter 1; erythroid K-Cl cotransporter 1; FLJ40489; Furosemide-sensitive K-Cl cotransporter; hKCC1; Kcc1; K-Cl cotransporter; K-Cl cotransporter 1; K-Cl cotransporter 4; K-Cl cotransporter KCC1; K-Cl Co-transporter-1; mKCC1; potassium chloride cotransporter SLC12A4; potassium/chloride cotransporter 1; potassium-chloride cotransporter-1; RBCKCC1; RBKCC1; rKCC1; Slc12a4; solute carrier family 12 (potassium/chloride transporter), member 4; solute carrier family 12 (potassium/chloride transporters), member 4; solute carrier family 12 member 4; solute carrier family 12, member 4
Gene Symbols SLC12A4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC12A4 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6560
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.