Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCC2 Monoclonal Antibody (N1/12), FITC
GREENER_CHOICE

Catalog No. PIMA545496
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545496 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545496 Supplier Invitrogen™ Supplier No. MA545496
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45496 was sufficient for detection of KCC2 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 140kDa. This antibody was formerly sold as clone S1-12.

Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCC2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N1/12
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene SLC12A5
Gene Accession No. Q63633, Q91V14, Q9H2X9
Gene Alias EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5
Gene Symbols SLC12A5
Host Species Mouse
Immunogen Fusion protein amino acids 932-1043 corresponding to rat KCC2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171373, 57138, 57468
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.