Learn More
Invitrogen™ KCC2 Monoclonal Antibody (N1/12)

Description
1 μg/mL of MA5-27610 was sufficient for detection of KCC2 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 140kDa. This antibody was formerly sold as clone S1-12.
Specifications
Specifications
| Antigen | KCC2 |
| Applications | Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | N1/12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene | SLC12A5 |
| Gene Accession No. | Q63633, Q91V14, Q9H2X9 |
| Gene Alias | EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.