Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCC2/SLC12A5 Antibody (905705), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8369E
Description
KCC2/SLC12A5 Monoclonal antibody specifically detects KCC2/SLC12A5 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
KCC2/SLC12A5 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
905705 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5 | |
NS0 mouse myeloma cell line transfected with human SLC12A5, Accession # Q9H2X9 | |
0.1 mL | |
Potassium Channels | |
57468 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction