Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCC2/SLC12A5 Antibody (S1-12), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$560.50
Specifications
Antigen | KCC2/SLC12A5 |
---|---|
Clone | S1-12 |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
KCC2/SLC12A5 Monoclonal specifically detects KCC2/SLC12A5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
KCC2/SLC12A5 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
57468 | |
Fusion protein amino acids 932-1043 corresponding to rat KCC2 | |
Primary | |
Detects 140kDa. |
S1-12 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Potassium Channels | |
Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5 | |
SLC12A5 | |
IgG2a | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title