Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCC3/SLC12A6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095618 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095618 20 μL
NB095617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095618 Supplier Novus Biologicals Supplier No. NBP30377220UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCC3/SLC12A6 Polyclonal specifically detects KCC3/SLC12A6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen KCC3/SLC12A6
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ACCPN, agenesis of corpus callosum and peripheral neuropathy (Andermann syndrome), DKFZp434D2135, EC 1.2.4.1, EC 6.1.1.3, Electroneutral potassium-chloride cotransporter 3, KCC3A, KCC3B, KCC3potassium chloride cotransporter 3, K-Cl cotransporter 3, potassium chloride cotransporter KCC3a-S3, potassium-chloride transporter-3a, potassium-chloride transporter-3b, solute carrier family 12 (potassium/chloride transporters), member 6, solute carrier family 12 member 6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-190 of human KCC3/SLC12A6 (NP_598408.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9990
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.