Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588472
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588472 Supplier Invitrogen™ Supplier No. PA588472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPTNFTVVPV EAHADGGGDE TAERTEAPGT PEGPEPERPS PGDGNPRENS PFLNNVEVEQ ESFFEGKNMA; Positive Samples: Mouse kidney, Mouse liver, Rat kidney; Cellular Location: Cell membrane, Multi-pass membrane protein.

Mediates electroneutral potassium-chloride cotransport when activated by cell swelling. May mediate K(+) uptake into Deiters' cells in the cochlea and contribute to K(+) recycling in the inner ear. Important for the survival of cochlear outer and inner hair cells and the maintenance of the organ of Corti. May be required for basolateral Cl(-) extrusion in the kidney and contribute to renal acidification. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCC4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene SLC12A7
Gene Accession No. Q5RK27, Q9WVL3
Gene Alias AA408796; D13Ertd261e; electroneutral potassium-chloride cotransporter 4; EMBL:AAH86339.1, ECO:0000312; furosemide-sensitive KCl cotransporter 4; Kcc4; K-Cl cotransporter 4; K-Cl cotransporter KCC4; potassium/chloride transporter KCC4; RGD:1359672}; Slc12a7; slc12a7 {ECO:0000312; solute carrier family 12 (potassium/chloride transporter), member 7; solute carrier family 12 (potassium/chloride transporters), member 7; solute carrier family 12 member 7; solute carrier family 12, member 7
Gene Symbols SLC12A7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human SLC12A7 (NP_0065892).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20499, 308069
Target Species Mouse, Rat, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.