Learn More
KCC4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304442A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Transcriptional activator. Binds specifically to the DNA sequence 5′-AACAAT-3′. Plays a key role in several developmental processes, including neurogenesis and skeleton formation.Specifications
KCC4 | |
Polyclonal | |
Unconjugated | |
SLC12A7 | |
KCC4, K-Cl cotransporter 4, electroneutral potassium-chloride cotransporter 4, potassium/chloride transporter KCC4, solute carrier family 12 member 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10723 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9Y666 | |
SLC12A7 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.