Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-KCC4/SLC12A7, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP185133
Description
KCC4/SLC12A7 Polyclonal antibody specifically detects KCC4/SLC12A7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KCC4/SLC12A7 | |
Unconjugated | |
Affinity Purified | |
DKFZp434F076, KCC4Electroneutral potassium-chloride cotransporter 4, K-Cl cotransporter 4, potassium/chloride transporter KCC4, solute carrier family 12 (potassium/chloride transporters), member 7, solute carrier family 12 member 7 | |
SLC12A7 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:AARTQAPPTPDKVQMTWTREKLIAEKYRSRDTSLSGFKDLFSMKPDQSNV | |
Polyclonal | |
Immunogen affinity purified | |
RUO | |
Human,Mouse,Rat |
Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin) | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
10723 | |
Rabbit | |
IgG | |
Primary | |
0.1 ml | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Specificity of human KCC4/SLC12A7 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only