Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCC4/SLC12A7 Antibody (891526), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB9030PECY55
Description
KCC4/SLC12A7 Monoclonal antibody specifically detects KCC4/SLC12A7 in Human samples. It is validated for Flow CytometrySpecifications
| KCC4/SLC12A7 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10723 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 891526 | |
| Flow Cytometry | |
| DKFZp434F076, KCC4Electroneutral potassium-chloride cotransporter 4, K-Cl cotransporter 4, potassium/chloride transporter KCC4, solute carrier family 12 (potassium/chloride transporters), member 7, solute carrier family 12 member 7 | |
| NS0 mouse myeloma cell line transfected with human SLC12A7, Met1-Ser1083, Accession # Q9Y666 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction