Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KChIP2 Antibody (OTI3C4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245471
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
KChIP2 Monoclonal specifically detects KChIP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
KChIP2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry | |
A-type potassium channel modulatory protein 2, cardiac voltage gated potassium channel modulatory subunit, Cardiac voltage-gated potassium channel modulatory subunit, DKFZp566L1246, KChIP2, KCHIP2Kv channel-interacting protein 2, Kv channel interacting protein 2, MGC17241, potassium channel interacting protein 2, Potassium channel-interacting protein 2 | |
Mouse | |
28.8 kDa | |
0.1 mL | |
Vision | |
30819 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3C4 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
KCNIP2 | |
Full length human recombinant protein of human KCNIP2 (NP_775284) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction