Learn More
Invitrogen™ KChIP2 Monoclonal Antibody (OTI3D8)

Description
KChIP2 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | KChIP2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3D8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | KCNIP2 |
| Gene Accession No. | Q9NS61 |
| Gene Alias | A-type potassium channel modulatory protein 2; Cardiac voltage-gated potassium channel modulatory subunit; DKFZp566L1246; KCHIP2; KChlP2; Kcnip2; Kv channel interacting protein 2; Kv channel-interacting protein; Kv channel-interacting protein 2; MGC17241; potassium channel auxiliary subunit KCHIP2; potassium channel-interacting protein 2; potassium voltage-gated channel interacting protein 2; si:ch73-173h19.2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.