Learn More
Invitrogen™ KCMF1 Polyclonal Antibody

Description
Peptide sequence: SVEQPQSFTC PYCGKMGYTE TSLQEHVTSE HAETSTEVIC PICAALPGGD Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 93%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 93%.
Specifications
Specifications
| Antigen | KCMF1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | KCMF1 |
| Gene Accession No. | Q9P0J7 |
| Gene Alias | 1700094M07Rik; clone DEBT-91; DEBT91; debt-91; Differentially expressed in branching tubulogenesis 91; DKFZp434L1021; E3 ubiquitin-protein ligase KCMF1; FGF-induced in gastric cancer; FGF-induced ubiquitin-protein ligase in gastric cancers; FIGC; kcmf1; kcmf1 protein; PCMF; Pmcf; Potassium channel modulatory factor; potassium channel modulatory factor 1; potassium channel modulatory factor DEBT-91; RING-type E3 ubiquitin transferase KCMF1; si:ch211-220b11.7; zgc:64160; zinc finger, ZZ domain containing 1; zKCMF1; ZZ-type zinc finger-containing protein 1; ZZZ1 |
| Gene Symbols | KCMF1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.