Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCMF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113665 Supplier Invitrogen™ Supplier No. PA5113665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STLVREESSS SDEDDRGEMA DFGAMGCVDI MPLDVALENL NLKESNKGNE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

KCMF1 belongs to the KCMF1 family. It contains 1 C2H2-type zinc finger and 1 ZZ-type zinc finger. It has intrinsic E3 ubiquitin ligase activity and promotes ubiquitination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCMF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCMF1
Gene Accession No. Q9P0J7
Gene Alias 1700094M07Rik; clone DEBT-91; DEBT91; debt-91; Differentially expressed in branching tubulogenesis 91; DKFZp434L1021; E3 ubiquitin-protein ligase KCMF1; FGF-induced in gastric cancer; FGF-induced ubiquitin-protein ligase in gastric cancers; FIGC; kcmf1; kcmf1 protein; PCMF; Pmcf; Potassium channel modulatory factor; potassium channel modulatory factor 1; potassium channel modulatory factor DEBT-91; RING-type E3 ubiquitin transferase KCMF1; si:ch211-220b11.7; zgc:64160; zinc finger, ZZ domain containing 1; zKCMF1; ZZ-type zinc finger-containing protein 1; ZZZ1
Gene Symbols KCMF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human KCMF1 (aa 326-375).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56888
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.