Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcna2 Mouse anti-Mouse, Rat, PerCP, Clone: C25, Abnova™

Catalog No. 89338903 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-903 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-903 Supplier Abnova Corporation Supplier No. MAB19399

Mouse Monoclonal Antibody.

Kcna2 Monoclonal antibody specifically detects Kcna2 in Mouse, Rat samples. It is validated for WB, IHC-P.

Specifications

Antigen Kcna2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C25
Conjugate PerCP
Description potassium voltage-gated channel, shaker-related subfamily, member 2
Gene Kcna2
Gene Alias Akr6a4, Kca1-2, Kv1.2, Mk-2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 428-499 at C-terminus of rat Kcna2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 16490
Target Species Mouse, Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.