Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcna4 Mouse anti-Mouse, Rat, FITC, Clone:H8, Abnova™

Catalog No. 89334155 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-155 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-155 Supplier Abnova Corporation Supplier No. MAB17437

Mouse monoclonal antibody raised against synthetic peptide of rat Kcna4.

Specifications

Antigen Kcna4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone H8
Conjugate FITC
Description potassium voltage-gated channel, shaker-related subfamily, member 4
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Kcna4
Gene Alias KCHAN, Kv1.4, Kv4, RHK1, RK3
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 13-37 of rat Kcna4.
Quantity 100 μg
Gene ID (Entrez) 25469
Target Species Mouse, Rat
Content And Storage Store at 4°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.