Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcna5 Mouse anti-Rat, ATTO 488, Clone: D23, Abnova™

Catalog No. 89338870 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-870 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-870 Supplier Abnova Corporation Supplier No. MAB19366

Mouse Monoclonal Antibody.

Kcna5 Monoclonal antibody specifically detects Kcna5 in Rat samples. It is validated for WB, ICC, IHC-P, IF.

Specifications

Antigen Kcna5
Applications Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone D23
Conjugate Atto 488
Description potassium voltage-gated channel, shaker-related subfamily, member 5
Gene Kcna5
Gene Alias Kv1, Kv1.5
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 542-602 at C-terminus of rat Kcna5
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 25470
Target Species Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.