Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcna5, Mouse anti-Rat, Clone: D23, Abnova™

Catalog No. 89999481 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-481 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-481 Supplier Abnova Corporation Supplier No. MAB16498

Mouse Monoclonal Antibody

Specifications

Antigen Kcna5
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone D23
Conjugate Unconjugated
Dilution Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Gene potassium voltage-gated channel, shaker-related subfamily, member 5
Gene Alias Kv1/Kv1.5
Gene Symbols Kcna5
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 542-602 at C-terminus of rat Kcna5.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25470
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.