Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kcna6, Mouse anti-Rat, Biotin, Clone: AH11, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16509
Description
Sequence: RERRSSYLPTPHRAYAEKRSpecifications
Kcna6 | |
Monoclonal | |
Biotin | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
Kv1.6 | |
Mouse | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG3 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
AH11 | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) The optimal working dilution should be determined by the end user. | |
potassium voltage gated channel, shaker related subfamily, member 6 | |
Kcna6 | |
A synthetic peptide corresponding to amino acids 507-525 at C-terminus of rat Kcna6. | |
RUO | |
64358 | |
Store at 4°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction