Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNAB1 Monoclonal Antibody (OTI2A8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503965

Description
KCNAB1 is a protein involved in potassium channels representing the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNAB1 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily.Specifications
KCNAB1 | |
Monoclonal | |
0.52 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q14722 | |
KCNAB1 | |
Full length human recombit protein of human KCNAB1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI2A8 | |
Unconjugated | |
KCNAB1 | |
AKR6A3, KCNA1B, KV-BETA-1, Kvb1.3, hKvBeta3, hKvb3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7881 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction