Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNAB1 Monoclonal Antibody (OTI2F1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503970S


Description
KCNAB1 Monoclonal antibody specifically detects KCNAB1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
KCNAB1 | |
Monoclonal | |
0.96 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q14722 | |
Mouse | |
Affinity chromatography | |
RUO | |
7881 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2F1 | |
Unconjugated | |
KCNAB1 | |
AKR6A3; Akr8a8; hKvb3; hKvBeta3; K(+) channel subunit beta-1; K+ channel Beta1a chain; KCNA1B; KCNAB1; Kvb1; Kvb1.3; KVBETA1; Kv-beta-1; Kvbeta1.1; KvBeta3; mKv(beta)1; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel subunit Kv beta 1; potassium channel subunit Kv beta 1.1; potassium channel subunit Kv beta 1.2; potassium channel subunit Kv beta 1.3; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium channel, voltage-gated shaker related subfamily A regulatory beta subunit 1; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 1; potassium voltage-gated channel shaker-related subfamily beta member 1; potassium voltage-gated channel subfamily A member regulatory beta subunit 1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; voltage-gated potassium channel beta subunit; voltage-gated potassium channel subunit beta-1 | |
Full length human recombinant protein of human KCNAB1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction