Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNAB1 Monoclonal Antibody (OTI9F7), TrueMAB™, OriGene
SDP

Catalog No. 501674740
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-4740 100 μL
50-254-0600 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-4740 Supplier OriGene Supplier No. TA503967
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCNAB1 is a protein involved in potassium channels representing the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNAB1 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily.

Specifications

Antigen KCNAB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI9F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KCNAB1
Gene Accession No. Q14722
Gene Alias AKR6A3, KCNA1B, KV-BETA-1, Kvb1.3, hKvBeta3, hKvb3
Gene Symbols KCNAB1
Host Species Mouse
Immunogen Full length human recombit protein of human KCNAB1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7881
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.