Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNAB1 Monoclonal Antibody (OTI9F7), TrueMAB™, OriGene
SDP

Catalog No. 502540600
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0600 30 μL
50-167-4740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0600 Supplier OriGene Supplier No. TA503967S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCNAB1 Monoclonal antibody specifically detects KCNAB1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen KCNAB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI9F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KCNAB1
Gene Accession No. Q14722
Gene Alias AKR6A3; Akr8a8; hKvb3; hKvBeta3; K(+) channel subunit beta-1; K+ channel Beta1a chain; KCNA1B; KCNAB1; Kvb1; Kvb1.3; KVBETA1; Kv-beta-1; Kvbeta1.1; KvBeta3; mKv(beta)1; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel subunit Kv beta 1; potassium channel subunit Kv beta 1.1; potassium channel subunit Kv beta 1.2; potassium channel subunit Kv beta 1.3; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium channel, voltage-gated shaker related subfamily A regulatory beta subunit 1; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 1; potassium voltage-gated channel shaker-related subfamily beta member 1; potassium voltage-gated channel subfamily A member regulatory beta subunit 1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; voltage-gated potassium channel beta subunit; voltage-gated potassium channel subunit beta-1
Host Species Mouse
Immunogen Full length human recombinant protein of human KCNAB1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7881
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.