Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnab1 Mouse anti-Mouse, Rat, Alkaline Phosphatase, Clone: XF1, Abnova™

Catalog No. 89338889 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-889 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-889 Supplier Abnova Corporation Supplier No. MAB19385

Mouse Monoclonal Antibody.

Kcnab1 Monoclonal antibody specifically detects Kcnab1 in Mouse, Rat samples. It is validated for WB-Ti, IHC-P.

Specifications

Antigen Kcnab1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone XF1
Conjugate Alkaline Phosphatase (AP)
Description potassium voltage-gated channel, shaker-related subfamily, beta member 1
Gene Kcnab1
Gene Alias Akr8a8, Kvbeta1.1, mKv(beta)1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 7-28 at N-terminus of rat Kcnab1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 16497
Target Species Mouse, Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.