Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kcnab1, Mouse anti-Rat, Clone: A3, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16512
Description
Sequence: ADIPSPKLGLPKSSESALKCSpecifications
Kcnab1 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
KvBeta3/MGC105463 | |
Mouse | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
A3 | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
potassium voltage-gated channel, shaker-related subfamily, beta member 1 | |
Kcnab1 | |
A synthetic peptide corresponding to amino acids 9-28 at N-terminus of rat Kcnab1.2. | |
RUO | |
29737 | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction