Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kcnab2 Mouse anti-Rat, APC, Clone:G6, Abnova™
Mouse monoclonal antibody raised against full length recombinant rat Kcnab2.
Supplier: Abnova Corporation MAB17444
Specifications
Kcnab2 | |
Monoclonal | |
APC | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
Kcnab2 | |
Recombinant protein corresponding to full length rat Kcnab2. | |
29738 | |
Store at 4°C. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
G6 | |
potassium voltage-gated channel, shaker-related subfamily, beta member 2 | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
Mouse | |
100 μg | |
Rat | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction