Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnab2, Mouse anti-Rat, Biotin, Clone: G6, Abnova™

Catalog No. 89999448 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-448 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-448 Supplier Abnova Corporation Supplier No. MAB16507

Mouse Monoclonal Antibody

Specifications

Antigen Kcnab2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone G6
Conjugate Biotin
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Gene potassium voltage-gated channel, shaker-related subfamily, beta member 2
Gene Alias -
Gene Symbols Kcnab2
Host Species Mouse
Immunogen Recombinant protein corresponding to full length rat Kcnab2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29738
Target Species Rat
Content And Storage Store at 4°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.