Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNAB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | KCNAB2 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1857
![]() ![]() |
Proteintech Group Inc
178901AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Kcnab2 is a voltage-gated potassium channels (KV) belong to the 6-TM family of potassium channel that also comprises the Ca2+-activated Slo (actually 7-TM) and the Ca2+-activated SK subfamilies. The alpha-subunits contain a single pore-forming region and combine to form tetramers. Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of potassium channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). Kcnab2 is one of the beta subunits, which are auxiliary proteins associating with functional Kv-alpha subunits. Alternative splicing of the Kcnab2 gene results in multiple transcript variants encoding distinct isoforms. Diseases associated with KCNAB2 include Chromosome 1P36 Deletion Syndrome.Specifications
KCNAB2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P62482, P62483, Q13303 | |
16498, 29738, 8514 | |
KCNAB2 Fusion Protein Ag12096 | |
Primary | |
-20°C | |
KCNAB2 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AKR6A5, HKvbeta2, HKvbeta2.1, HKvbeta2.2, K(+) channel subunit beta 2, KCNA2B, KCNAB2, KCNK2, KV BETA 2, TREK, TREK1 | |
KCNAB2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title