Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNAB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539531
Description
Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. The KV gene family encodes more than 30 proteins that comprise the subunits of the K+ channels, and they vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha subunits (KV), which include the KV1, KV2, KV3 and KV4 proteins, and accessory or KV-subunits that modify the gating properties of the coexpressed KV subunits. KV beta.3 is an accessory K+ channel protein which regulates the activity of the pore-forming alpha subunit and alters the functional properties of Kv1.5. KV beta.3 localizes to the cytoplasm and is expressed in the brain, with highest expression detected in the cerebellum, and weakest expression seen in the frontal and temporal lobes. No KV beta.3 expression is detected in the heart, spinal cord, lung, liver, kidney, pancreas, placenta or skeletal muscle.
Specifications
| KCNAB3 | |
| Polyclonal | |
| Unconjugated | |
| KCNAB3 | |
| Voltage-gated potassium channel subunit beta-3, K(+) channel subunit beta-3, Kv-beta-3, KCNAB3, KCNA3B, KV-beta.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16499, 58981, 9196 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43448, P97382, Q63494 | |
| KCNAB3 | |
| A synthetic peptide derived from the C-terminal region of human KCNAB3 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction