Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNAB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567484
Description
Immunogen sequence: CGLITSKYDGR VPDTCRASIK GYQWLKDKVQ SEDGKKQQAK VMDLLPVAHQ Highest antigen sequence identity to the following orthologs - mouse 84%, rat 80%.
Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. The KV gene family encodes more than 30 proteins that comprise the subunits of the K+ channels, and they vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha subunits (KV), which include the KV1, KV2, KV3 and KV4 proteins, and accessory or KV-subunits that modify the gating properties of the coexpressed KV subunits. KV beta.3 is an accessory K+ channel protein which regulates the activity of the pore-forming alpha subunit and alters the functional properties of Kv1.5. KV beta.3 localizes to the cytoplasm and is expressed in the brain, with highest expression detected in the cerebellum, and weakest expression seen in the frontal and temporal lobes. No KV beta.3 expression is detected in the heart, spinal cord, lung, liver, kidney, pancreas, placenta or skeletal muscle.
Specifications
KCNAB3 | |
Polyclonal | |
Unconjugated | |
KCNAB3 | |
AKR6A9; C330022D06Rik; Ckbeta3; K(+) channel subunit beta-3; KCNA3.1B; KCNA3B; Kcnab3; Kcnab4; KV-beta.3; Kv-beta-3; mKv(beta)4; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 3; potassium channel, voltage gated subfamily A regulatory beta subunit 3; potassium channel, voltage-dependent, beta-3 subunit; potassium channel, voltage-gated shaker-related subfamily A regulatory beta subunit 3; potassium voltage gated channel, shaker related subfamily, beta member 3; potassium voltage-gated channel subfamily A regulatory beta subunit 3; potassium voltage-gated channel, shaker-related subfamily, beta member 3; RCK beta3; Voltage-gated potassium channel subunit beta-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9196 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43448 | |
KCNAB3 | |
Recombinant Human KCNAB3. Recombinant protein control fragment (Product #RP-108137). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction