Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNAB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536970 Supplier Thermo Scientific Supplier No. PA536970
Add to Cart
Add to Cart

KCNAB3 Polyclonal Antibody

KCNAB3 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNAB3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNAB3
Gene Accession No. O43448, P97382, Q63494
Gene Alias AKR6A9; C330022D06Rik; Ckbeta3; K(+) channel subunit beta-3; KCNA3.1B; KCNA3B; Kcnab3; Kcnab4; KV-beta.3; Kv-beta-3; mKv(beta)4; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 3; potassium channel, voltage gated subfamily A regulatory beta subunit 3; potassium channel, voltage-dependent, beta-3 subunit; potassium channel, voltage-gated shaker-related subfamily A regulatory beta subunit 3; potassium voltage gated channel, shaker related subfamily, beta member 3; potassium voltage-gated channel subfamily A regulatory beta subunit 3; potassium voltage-gated channel, shaker-related subfamily, beta member 3; RCK beta3; Voltage-gated potassium channel subunit beta-3
Gene Symbols KCNAB3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 337-382 of human KV beta3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16499, 58981, 9196
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.