Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114305 Supplier Invitrogen™ Supplier No. PA5114305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNB1 Polyclonal Antibody for Flow, IHC (P), Western Blot

Mediates the voltage-dependent potassium ion permeability of excitable membranes. Channels open or close in response to the voltage difference across the membrane, letting potassium ions pass in accordance with their electrochemical gradient.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNB1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Kcnb1
Gene Accession No. P15387, Q03717, Q14721
Gene Alias Delayed rectifier potassium channel 1; delayed rectifier potassium channel Kv2.1; DRK1; DRK1PC; KCB1; Kcnb1; Kcr1-1; Kv2.1; mShab; potassium channel Kv2.1; potassium channel, voltage gated Shab-related subfamily B, member 1; potassium voltage gated channel, Shab-related subfamily, member 1; potassium voltage-gated channel subfamily B member 1; potassium voltage-gated channel, Shab-related subfamily, member 1; Shab; voltage-gated potassium channel subunit Kv2.1
Gene Symbols Kcnb1
Host Species Rabbit
Immunogen Synthetic peptide within rat KCNB1 aa 808-857.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16500, 25736, 3745
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.