Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986539
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6539 Supplier Bioss Supplier No. BS5402R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.

Specifications

Antigen KCNC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KCNC1
Gene Accession No. P48547
Gene Alias BOS_14495; C230009H10Rik; EPM7; FLJ41162; FLJ42249; FLJ43491; KCNC1; Kcr2-1; Krc2 1; KShIIIB; Kv3.1; KV4; MGC129855; NGK2; NGK2-KV4; Potassium channel gene 1 (alternative splicing product described in Luneau et al 1991); potassium channel, voltage gated Shaw related subfamily C, member 1; potassium channel, voltage gated Shaw-related subfamily C, member 1; potassium voltage gated channel, Shaw-related subfamily, member 1; Potassium voltage-gated channel subfamily C member 1; potassium voltage-gated channel, Shaw-related subfamily, member 1; RAW2; Shaw; Shaw-related voltage-gated potassium channel protein 1; voltage-gated potassium channel KCNC1; voltage-gated potassium channel protein KV3.1; Voltage-gated potassium channel subunit Kv3.1; Voltage-gated potassium channel subunit Kv4
Gene Symbols KCNC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KCNC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3746
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.