Learn More
KV3.2 (KCNC2) Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA554050
Description
Immunogen sequence: SQIQEPCCSD LFLFPDESGN VSQESGPTYA SFSHHFISDA MTGVPTENDD FCILFAPKAA MQEKEEEPVI KIMVDDAIVI RDNYFSLPVN KTDTSKAPLH FPIPVIRYVV KEVSLVWHLY GGKDFGTVPP TSPA Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat
Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. The KV gene family encodes more than 30 proteins that comprise the subunits of the K+ channels. The subunits vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha subunits, which include the KV1, KV2, KV3, KV4 and KV9 proteins, as well as accessory subunits that modify the gating properties of the coexpressed KV subunits. KV3.2 is a multipass membrane protein that regulates the voltage-dependent K+ permeability of excitable membranes. The tail of KV3.2 may be influential in the targeting of the channel to specific subcellular compartments and/or the regulation of channel activity.
Specifications
KV3.2 (KCNC2) | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PR1 | |
KCNC2 | |
Recombit protein corresponding to Human KCNC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
KCNC2 | |
Potassium voltage-gated channel subfamily C member 2, Voltage-gated potassium channel Kv3.2, KCNC2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3747 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only