Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ KCND3 (Human) Recombinant Protein
Click to view available options
Quantity:
10 μg
Description
Human KCND3 full-length ORF (ADR83090.1) recombinant protein without tag.
Specifications
Specifications
For Use With (Application) | Antibody Production |
Gene ID (Entrez) | 3752 |
Name | potassium voltage-gated channel, Shal-related subfamily, member 3 |
Quantity | 10 μg |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Gene Alias | KCND3L, KCND3S, KSHIVB, KV4.3, MGC142035, MGC142037 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction