Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnd3 Mouse anti-Rat, ATTO 390, Clone: Y19, Abnova™

Catalog No. 89339012 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-012 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-012 Supplier Abnova Corporation Supplier No. MAB19727

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody.

Kcnd3 Monoclonal antibody specifically detects Kcnd3 in Rat samples. It is validated for WB, IHC-P.

Specifications

Antigen Kcnd3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Y19
Conjugate Atto 390
Description potassium voltage gated channel, Shal-related family, member 3
Gene Kcnd3
Gene Alias Kv4.3
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 415-636 at C-terminus of rat Kcnd3
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 65195
Target Species Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.